3-(Thien-2-yl)benzylamine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(3-Thien-2-ylphenyl)methylamine
OR9262-1G
OR9262-250MG
APOSOR9262-1GEA
306
EUR
APOSOR9262-1G
APOSOR9262-250MG
3-(Thien-2-yl)benzylamine 97%
3-(Thien-2-yl)benzylamine
Formula:
C₁₁H₁₁NS MW: 189,277 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08435868 CAS Number: 859850-86-5 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...