Methyl 4-methylthiazole-2-carboxylate 95%
Supplier: Apollo Scientific
OR926326-1G
OR926326-5G
APOSOR926326-1GEA
85.7
EUR
APOSOR926326-1G
APOSOR926326-5G
Methyl 4-methylthiazole-2-carboxylate 95%
Methyl 4-methylthiazole-2-carboxylate
Formula:
C₆H₇NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD08275081 CAS Number: 14542-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...