7-Methoxy-3,4-dihydroquinolin-2(1H)-one 95%
Supplier: Apollo Scientific
OR926381-1G
OR926381-250MG
APOSOR926381-1GEA
53.9
EUR
APOSOR926381-1G
APOSOR926381-250MG
7-Methoxy-3,4-dihydroquinolin-2(1H)-one 95%
7-Methoxy-3,4-dihydroquinolin-2(1H)-one
Formula:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15144280 CAS Number: 22246-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...