6-Ethyl-4-hydroxycoumarin 95%

Supplier: Apollo Scientific

OR926382-1G OR926382-5G
APOSOR926382-1GEA 168 EUR
APOSOR926382-1G APOSOR926382-5G
6-Ethyl-4-hydroxycoumarin 95%
6-Ethyl-4-hydroxycoumarin
Formula: C₁₁H₁₀O₃
MW: 190,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424528
CAS Number: 55005-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR