4-Methylthiophene-2-sulphonyl chloride 95%

Supplier: Apollo Scientific

OR926528-100MG OR926528-1G OR926528-250MG
APOSOR926528-100MGEA 147 EUR
APOSOR926528-100MG APOSOR926528-1G APOSOR926528-250MG
4-Methylthiophene-2-sulphonyl chloride 95%
4-Methylthiophene-2-sulphonyl chloride
Formula: C₅H₅ClO₂S₂
MW: 196,675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864497
CAS Number: 69815-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR