1,2,3,4-Tetrahydro-1-methylquinoline 95%
Supplier: Apollo Scientific
Synonyms:
1-Methyl-1,2,3,4-tetrahydroquinoline
OR926569-10G
OR926569-1G
OR926569-5G
APOSOR926569-10GEA
481
EUR
APOSOR926569-10G
APOSOR926569-1G
APOSOR926569-5G
1,2,3,4-Tetrahydro-1-methylquinoline 95%
1,2,3,4-Tetrahydro-1-methylquinoline
Formula:
C₁₀H₁₃N MW: 147,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00130001 CAS Number: 491-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...