5,6,7,8-Tetrahydroquinazoline 95%

Supplier: Apollo Scientific

OR926731-1G
APOSOR926731-1GEA 353 EUR
APOSOR926731-1G
5,6,7,8-Tetrahydroquinazoline 95%
5,6,7,8-Tetrahydroquinazoline
Formula: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15146638
CAS Number: 5632-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR