1-Vinyl-1H-pyrazole-4-carboxylic acid 95%
Supplier: Apollo Scientific
OR926841-1G
OR926841-250MG
APOSOR926841-1GEA
865
EUR
APOSOR926841-1G
APOSOR926841-250MG
1-Vinyl-1H-pyrazole-4-carboxylic acid 95%
1-Vinyl-1H-pyrazole-4-carboxylic acid
Formula:
C₆H₆N₂O₂ MW: 138,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198429 CAS Number: 905307-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...