1-Cyclopropyl-1H-pyrazole 95%

Supplier: Apollo Scientific

OR926889-1G OR926889-250MG
APOSOR926889-1GEA 735 EUR
APOSOR926889-1G APOSOR926889-250MG
1-Cyclopropyl-1H-pyrazole 95%
1-Cyclopropyl-1H-pyrazole
Formula: C₆H₈N₂
MW: 108,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22548191
CAS Number: 1151814-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR