(4-Amino-3-methoxyphenyl)methanol 95+%

Supplier: Apollo Scientific
Warning

OR926918-1G OR926918-250MG OR926918-5G
APOSOR926918-1GEA 142 EUR
APOSOR926918-1G APOSOR926918-250MG APOSOR926918-5G
(4-Amino-3-methoxyphenyl)methanol 95+%
(4-Amino-3-methoxyphenyl)methanol
Formula: C₈H₁₁NO₂
MW: 153,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236863
CAS Number: 148459-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR