tert-Butyl cyclopropanecarboxylate 95%

Supplier: Apollo Scientific

OR927117-1G OR927117-25G OR927117-5G
APOSOR927117-1GEA 24.4 EUR
APOSOR927117-1G APOSOR927117-25G APOSOR927117-5G
tert-Butyl cyclopropanecarboxylate 95%
tert-Butyl cyclopropanecarboxylate
Formula: C₈H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD09836696
CAS Number: 87661-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR