5-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride

Supplier: Apollo Scientific
Warning

OR927260-1G OR927260-5G OR927260-10G
APOSOR927260-1GEA 94.4 EUR
APOSOR927260-1G APOSOR927260-5G APOSOR927260-10G
5-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
5-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
Formula: C₅H₇ClN₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD24335262
CAS Number: 1434128-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR