1-(1,5-Dimethyl-1H-indazol-3-yl)methanamine 95%

Supplier: Apollo Scientific

OR927289-1G OR927289-250MG
APOSOR927289-1GEA 735 EUR
APOSOR927289-1G APOSOR927289-250MG
1-(1,5-Dimethyl-1H-indazol-3-yl)methanamine 95%
1-(1,5-Dimethyl-1H-indazol-3-yl)methanamine
Formula: C₁₀H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD11053913
CAS Number: 1060817-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR