6-Methylindolin-2-one 95%
Supplier: Apollo Scientific
OR927316-1G
OR927316-5G
APOSOR927316-1GEA
59.6
EUR
APOSOR927316-1G
APOSOR927316-5G
6-Methylindolin-2-one 95%
6-Methylindolin-2-one
Formula:
C₉H₉NO MW: 147,174 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08457972 CAS Number: 56341-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...