2-(Pyridin-2-yl)azepane 95%

Supplier: Apollo Scientific

OR927383-1G OR927383-250MG
APOSOR927383-1GEA 435 EUR
APOSOR927383-1G APOSOR927383-250MG
2-(Pyridin-2-yl)azepane 95%
2-(Pyridin-2-yl)azepane
Formula: C₁₁H₁₆N₂
MW: 176,258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663695
CAS Number: 383128-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR