5-Chloro-2-(methylsulphonyl)pyrimidine 96%

Supplier: Apollo Scientific
Warning

OR927453-1G
APOSOR927453-1GEA 306 EUR
APOSOR927453-1G
5-Chloro-2-(methylsulphonyl)pyrimidine 96%
5-Chloro-2-(methylsulphonyl)pyrimidine
Formula: C₅H₅ClN₂O₂S
MW: 192,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187881
CAS Number: 38275-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR