4-Acetoxyphenol 96%

Supplier: Apollo Scientific

OR927491-1G OR927491-5G
APOSOR927491-1GEA 31.6 EUR
APOSOR927491-1G APOSOR927491-5G
4-Acetoxyphenol 96%
4-Acetoxyphenol
Formula: C₈H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00799234
CAS Number: 3233-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR