4-[(Ethylamino)methyl]phenol 95%

Supplier: Apollo Scientific

OR927556-1G OR927556-5G
APOSOR927556-1GEA 357 EUR
APOSOR927556-1G APOSOR927556-5G
4-[(Ethylamino)methyl]phenol 95%
4-[(Ethylamino)methyl]phenol
Formula: C₉H₁₃NO
MW: 151,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09929168
CAS Number: 45966-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR