4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 95%
Supplier: Apollo Scientific
OR928099-1G
OR928099-250MG
APOSOR928099-1GEA
101
EUR
APOSOR928099-1G
APOSOR928099-250MG
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 95%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile
Formula:
C₈H₄ClN₃ MW: 177,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11046237 CAS Number: 920965-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...