4,5,6,7-Tetrahydro-1,2-benzisoxazole-3-carboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: 4,5,6,7-Tetrahydrobenzo[d]isoxazole-3-carboxylic acid

OR928142-1G OR928142-250MG
APOSOR928142-1GEA 437 EUR
APOSOR928142-1G APOSOR928142-250MG
4,5,6,7-Tetrahydro-1,2-benzisoxazole-3-carboxylic acid 95%
4,5,6,7-Tetrahydro-1,2-benzisoxazole-3-carboxylic acid
Formula: C₈H₉NO₃
MW: 167,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701438
CAS Number: 90005-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR