Ethyl 3-hydroxypicolinate 95%

Supplier: Apollo Scientific

OR928177-1G OR928177-250MG
APOSOR928177-1GEA 568 EUR
APOSOR928177-1G APOSOR928177-250MG
Ethyl 3-hydroxypicolinate 95%
Ethyl 3-hydroxypicolinate
Formula: C₈H₉NO₃
MW: 167,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656384
CAS Number: 73406-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR