1-(1-Ethylpyrrolidin-3-yl)-N-methylmethanamine 95%
Supplier: Apollo Scientific
OR928279-1G
OR928279-250MG
APOSOR928279-1GEA
864
EUR
APOSOR928279-1G
APOSOR928279-250MG
1-(1-Ethylpyrrolidin-3-yl)-N-methylmethanamine 95%
1-(1-Ethylpyrrolidin-3-yl)-N-methylmethanamine
Formula:
C₈H₁₈N₂ MW: 142,242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864495 CAS Number: 884504-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...