1-Methyl-3-(thien-2-yl)-1H-pyrazole-5-carboxaldehyde 97+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(5-Formyl-1-methyl-1H-pyrazol-3-yl)thiophene, 5-Formyl-1-methyl-3-(thien-2-yl)-1H-pyrazole
OR9283-1G
OR9283-250MG
APOSOR9283-1GEA
192
EUR
APOSOR9283-1G
APOSOR9283-250MG
1-Methyl-3-(thien-2-yl)-1H-pyrazole-5-carboxaldehyde 97+%
1-Methyl-3-(thien-2-yl)-1H-pyrazole-5-carboxaldehyde
Formula:
C₉H₈N₂OS MW: 192,238 g/mol Melting Pt: 76…77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08435884 CAS Number: 879896-48-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...