4-Chloro-2-methyl-5,6,7,8-tetrahydroquinazoline 95%

Supplier: Apollo Scientific
Warning

OR928510-1G OR928510-250MG
APOSOR928510-1GEA 275 EUR
APOSOR928510-1G APOSOR928510-250MG
4-Chloro-2-methyl-5,6,7,8-tetrahydroquinazoline 95%
4-Chloro-2-methyl-5,6,7,8-tetrahydroquinazoline
Formula: C₉H₁₁ClN₂
MW: 182,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755924
CAS Number: 90561-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR