N6,N6-Dimethyladenosine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2R,3R,4S,5R)-2-[6-(Dimethylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol, (2R,3R,4S,5R)-2-(6-(Dimethylamino)-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
OR928523-250MG
OR928523-1G
APOSOR928523-250MGEA
132
EUR
APOSOR928523-250MG
APOSOR928523-1G
N6,N6-Dimethyladenosine 97%
N6,N6-Dimethyladenosine
Formula:
C₁₂H₁₇N₅O₄ MW: 295,294 g/mol Melting Pt: 182…185 °C Storage Temperature: Freezer |
MDL Number:
MFCD00022822 CAS Number: 2620-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...