Methyl 5-(2-furyl)isoxazole-3-carboxylate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 5-(furan-2-yl)isoxazole-3-carboxylate

OR928555-5G
APOSOR928555-5GEA 266 EUR
APOSOR928555-5G
Methyl 5-(2-furyl)isoxazole-3-carboxylate 98%
Methyl 5-(2-furyl)isoxazole-3-carboxylate
Formula: C₉H₇NO₄
MW: 193,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03778977
CAS Number: 33545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR