1,3-Dihydro-2-benzofuran-4-amine 96%

Supplier: Apollo Scientific

Synonyms: 4-Amino-1,3-dihydroisobenzofuran

OR928572-1G OR928572-250MG
APOSOR928572-1GEA 149 EUR
APOSOR928572-1G APOSOR928572-250MG
1,3-Dihydro-2-benzofuran-4-amine 96%
1,3-Dihydro-2-benzofuran-4-amine
Formula: C₈H₉NO
MW: 135,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659158
CAS Number: 98475-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR