1-(3-Methyl-pyridin-2-ylmethyl)-piperazine 95%

Supplier: Apollo Scientific

OR928608-1G OR928608-250MG OR928608-5G
APOSOR928608-1GEA 241 EUR
APOSOR928608-1G APOSOR928608-250MG APOSOR928608-5G
1-(3-Methyl-pyridin-2-ylmethyl)-piperazine 95%
1-(3-Methyl-pyridin-2-ylmethyl)-piperazine
Formula: C₁₁H₁₇N₃
MW: 191,273 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05189259
CAS Number: 524673-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR