1,2,3,4-Tetrahydroquinoline-6-carbonitrile 96%

Supplier: Apollo Scientific

OR928761-100MG OR928761-1G OR928761-250MG
APOSOR928761-100MGEA 121 EUR
APOSOR928761-100MG APOSOR928761-1G APOSOR928761-250MG
1,2,3,4-Tetrahydroquinoline-6-carbonitrile 96%
1,2,3,4-Tetrahydroquinoline-6-carbonitrile
Formula: C₁₀H₁₀N₂
MW: 158,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14534464
CAS Number: 50741-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR