5-Bromo-1-methylpyrimidin-2(1H)-one 95%

Supplier: Apollo Scientific

OR928790-1G
APOSOR928790-1GEA 74.1 EUR
APOSOR928790-1G
5-Bromo-1-methylpyrimidin-2(1H)-one 95%
5-Bromo-1-methylpyrimidin-2(1H)-one
Formula: C₅H₅BrN₂O
MW: 189,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044136
CAS Number: 14248-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR