Butyl methanesulphonate 95%

Supplier: Apollo Scientific

OR928832-10G OR928832-5G OR928832-25G
APOSOR928832-10GEA 82.9 EUR
APOSOR928832-10G APOSOR928832-5G APOSOR928832-25G
Butyl methanesulphonate 95%
Butyl methanesulphonate
Formula: C₅H₁₂O₃S
MW: 152,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041257
CAS Number: 1912-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR