1-(3-Furyl)-N-methylethanamine 95%
Supplier: Apollo Scientific
OR928926-1G
OR928926-250MG
APOSOR928926-1GEA
720
EUR
APOSOR928926-1G
APOSOR928926-250MG
1-(3-Furyl)-N-methylethanamine 95%
1-(3-Furyl)-N-methylethanamine
Formula:
C₇H₁₁NO MW: 125,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04967012 CAS Number: 252372-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...