N-Fmoc-N-methyl-b-alanine 97%
Supplier: Apollo Scientific
Synonyms:
N-Fmoc-N-Methyl-beta-alanine, Fmoc-β-N-Me-Ala-OH
OR929329-1G
OR929329-500MG
OR929329-5G
APOSOR929329-1GEA
285
EUR
APOSOR929329-1G
APOSOR929329-500MG
APOSOR929329-5G
N-Fmoc-N-methyl-b-alanine 97%
N-Fmoc-N-methyl-b-alanine
Formula:
C₁₉H₁₉NO₄ MW: 325,358 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861327 CAS Number: 172965-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...