5-Pyridin-3-yl-2H-[1,2,4]triazol-3-ol 95%

Supplier: Apollo Scientific

OR929332-1G OR929332-250MG
APOSOR929332-1GEA 574 EUR
APOSOR929332-1G APOSOR929332-250MG
5-Pyridin-3-yl-2H-[1,2,4]triazol-3-ol 95%
5-Pyridin-3-yl-2H-[1,2,4]triazol-3-ol
Formula: C₇H₆N₄O
MW: 162,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441963
CAS Number: 59282-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR