4-Chloro-2-[(ethylamino)methyl]phenol 95%
Supplier: Apollo Scientific
OR929764-1G
OR929764-5G
APOSOR929764-1GEA
393
EUR
APOSOR929764-1G
APOSOR929764-5G
4-Chloro-2-[(ethylamino)methyl]phenol 95%
4-Chloro-2-[(ethylamino)methyl]phenol
Formula:
C₉H₁₂ClNO MW: 185,651 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09932367 CAS Number: 1016500-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...