4-Chloro-5,6,7,8-tetrahydroquinazoline 97%

Supplier: Apollo Scientific

OR929787-1G OR929787-5G
APOSOR929787-1GEA 243 EUR
APOSOR929787-1G APOSOR929787-5G
4-Chloro-5,6,7,8-tetrahydroquinazoline 97%
4-Chloro-5,6,7,8-tetrahydroquinazoline
Formula: C₈H₉ClN₂
MW: 168,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755923
CAS Number: 1125-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR