1-(2-Naphthyl)methanamine 95%
Supplier: Apollo Scientific
OR929803-1G
OR929803-250MG
APOSOR929803-1GEA
65.4
EUR
APOSOR929803-1G
APOSOR929803-250MG
1-(2-Naphthyl)methanamine 95%
1-(2-Naphthyl)methanamine
Formula:
C₁₁H₁₁N MW: 157,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01529867 CAS Number: 2018-90-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...