2-Bromo-3-methylpyridine 1-oxide 95%

Supplier: Apollo Scientific

OR929888-1G OR929888-250MG
APOSOR929888-1GEA 383 EUR
APOSOR929888-1G APOSOR929888-250MG
2-Bromo-3-methylpyridine 1-oxide 95%
2-Bromo-3-methylpyridine 1-oxide
Formula: C₆H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD20270297
CAS Number: 19230-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR