5,6,7,8-Tetrahydroquinolin-8-amine 95%
Supplier: Apollo Scientific
OR930035-1G
OR930035-250MG
APOSOR930035-1GEA
701
EUR
APOSOR930035-1G
APOSOR930035-250MG
5,6,7,8-Tetrahydroquinolin-8-amine 95%
5,6,7,8-Tetrahydroquinolin-8-amine
Formula:
C₉H₁₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD08234982 CAS Number: 298181-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...