N-(2-Hydroxypropyl)benzamide 95%

Supplier: Apollo Scientific

OR930200-1G OR930200-250MG
APOSOR930200-1GEA 865 EUR
APOSOR930200-1G APOSOR930200-250MG
N-(2-Hydroxypropyl)benzamide 95%
N-(2-Hydroxypropyl)benzamide
Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00464672
CAS Number: 23054-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR