4-Cyano-2-methoxybenzaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
4-Formyl-3-methoxybenzonitrile
OR930297-1G
OR930297-5G
APOSOR930297-1GEA
65.5
EUR
APOSOR930297-1G
APOSOR930297-5G
4-Cyano-2-methoxybenzaldehyde 95%
4-Cyano-2-methoxybenzaldehyde
Formula:
C₉H₇NO₂ MW: 161,157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07779491 CAS Number: 21962-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...