N-(2-Cyano-1-cyclopenten-1-yl)-acetamide 95%
Supplier: Apollo Scientific
OR930447-1G
OR930447-250MG
APOSOR930447-1GEA
437
EUR
APOSOR930447-1G
APOSOR930447-250MG
N-(2-Cyano-1-cyclopenten-1-yl)-acetamide 95%
N-(2-Cyano-1-cyclopenten-1-yl)-acetamide
Formula:
C₈H₁₀N₂O MW: 150,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01020954 CAS Number: 100377-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...