(2-Chloro-thiazol-5-ylmethyl)-methyl-amine 95%

Supplier: Apollo Scientific

OR930806-1G OR930806-250MG
APOSOR930806-1GEA 481 EUR
APOSOR930806-1G APOSOR930806-250MG
(2-Chloro-thiazol-5-ylmethyl)-methyl-amine 95%
(2-Chloro-thiazol-5-ylmethyl)-methyl-amine
Formula: C₅H₇ClN₂S
MW: 162,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18206726
CAS Number: 120740-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR