7-Chloro-DL-tryptophan 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-3-(7-chloro-1H-indol-3-yl)propanoic acid, H-DL-Trp(7-Cl)-OH

OR930930-250MG OR930930-500MG
APOSOR930930-250MGEA 172 EUR
APOSOR930930-250MG APOSOR930930-500MG
7-Chloro-DL-tryptophan 97%
7-Chloro-DL-tryptophan
Formula: C₁₁H₁₁ClN₂O₂
MW: 238,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264324
CAS Number: 153-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR