3-Chloro-5-nitrophenol 95%

Supplier: Apollo Scientific
Danger

OR930959-1G OR930959-5G
APOSOR930959-1GEA 219 EUR
APOSOR930959-1G APOSOR930959-5G
3-Chloro-5-nitrophenol 95%
3-Chloro-5-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173,554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15976952
CAS Number: 618-63-3
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR