Methyl 4-amino-5-chloro-2-methoxybenzoate 95%

Supplier: Apollo Scientific

OR931029-1G OR931029-250MG OR931029-5G
APOSOR931029-1GEA 149 EUR
APOSOR931029-1G APOSOR931029-250MG APOSOR931029-5G
Methyl 4-amino-5-chloro-2-methoxybenzoate 95%
Methyl 4-amino-5-chloro-2-methoxybenzoate
Formula: C₉H₁₀ClNO₃
MW: 215,634 g/mol
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD01034555
CAS Number: 20896-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR