1-Methyl-4-nitro-1H-indole 95%
Supplier: Apollo Scientific
OR931193-1G
OR931193-250MG
APOSOR931193-1GEA
270
EUR
APOSOR931193-1G
APOSOR931193-250MG
1-Methyl-4-nitro-1H-indole 95%
1-Methyl-4-nitro-1H-indole
Formula:
C₉H₈N₂O₂ MW: 176,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08236728 CAS Number: 91482-63-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...