6-Cyano-1H-indole-3-carboxylic acid 95%

Supplier: Apollo Scientific

OR931294-1G OR931294-250MG
APOSOR931294-1GEA 151 EUR
APOSOR931294-1G APOSOR931294-250MG
6-Cyano-1H-indole-3-carboxylic acid 95%
6-Cyano-1H-indole-3-carboxylic acid
Formula: C₁₀H₆N₂O₂
MW: 186.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954938
CAS Number: 174500-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR