5-Bromo-6-methylpicolinonitrile 95%
Supplier: Apollo Scientific
OR931501-1G
OR931501-250MG
APOSOR931501-1GEA
0
EUR
APOSOR931501-1G
APOSOR931501-250MG
5-Bromo-6-methylpicolinonitrile 95%
5-Bromo-6-methylpicolinonitrile
Formula:
C₇H₅BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD13181587 CAS Number: 1173897-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...