Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate 95%
Supplier: Apollo Scientific
OR932165-1G
OR932165-5G
APOSOR932165-1GEA
66.9
EUR
APOSOR932165-1G
APOSOR932165-5G
Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate 95%
Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate
Formula:
C₁₂H₁₁NO₃ MW: 217,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01314279 CAS Number: 52980-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...